Product Details
- SNP ID
-
rs10263802
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:138085889 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTCTTCATTTAAATGATCAGTTCC[A/G]GCCCATAAAATCTCAGTGATGTTTC
- Phenotype
-
MIM: 604741
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AKR1D1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75314391] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKR1D1
- Gene Name
- aldo-keto reductase family 1, member D1
View Full Product Details