Product Details
- SNP ID
-
rs7560763
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:84466147 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGTGAGAACGTGGCATTCTTTAC[C/T]TACCGTTATTTACTTAACTTCTTTA
- Phenotype
-
MIM: 603336
MIM: 611224
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAH6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76048397] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAH6
- Gene Name
- dynein axonemal heavy chain 6
- Gene
- SUCLG1
- Gene Name
- succinate-CoA ligase alpha subunit
There are no transcripts associated with this gene.
View Full Product Details