Product Details

SNP ID
rs9916782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40231788 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTCTTGGAAGTTGGGAGCTAGTA[A/G]TAATATGAGATATGGCTAGCTTAAA
Phenotype
MIM: 609692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WIPF2 PubMed Links

Gene Details

Gene
WIPF2
Gene Name
WAS/WASL interacting protein family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133264.4 Intron NP_573571.1
XM_005257083.2 Intron XP_005257140.1
XM_005257084.2 Intron XP_005257141.1
XM_011524412.1 Intron XP_011522714.1
XM_011524413.1 Intron XP_011522715.1

View Full Product Details