Product Details

SNP ID
rs7516131
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:40165599 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTGGGGGGACACTGTACATA[A/C]CTGTAGTTTTAACTCTTAATTTTAT
Phenotype
MIM: 180610
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RLF PubMed Links

Gene Details

Gene
RLF
Gene Name
rearranged L-myc fusion
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012421.3 Intron NP_036553.2
XM_017002016.1 Intron XP_016857505.1
XM_017002017.1 Intron XP_016857506.1
XM_017002018.1 Intron XP_016857507.1

View Full Product Details