Product Details

SNP ID
rs6925350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159738189 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCATTAATCAAGATACATATCTGA[A/G]CTATCACCATAAAGCTTCATCGAGT
Phenotype
MIM: 147460 MIM: 605442
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SOD2 PubMed Links

Gene Details

Gene
SOD2
Gene Name
superoxide dismutase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000636.3 Intron NP_000627.2
NM_001024465.2 Intron NP_001019636.1
NM_001024466.2 Intron NP_001019637.1
NM_001322814.1 Intron NP_001309743.1
NM_001322815.1 Intron NP_001309744.1
NM_001322816.1 Intron NP_001309745.1
NM_001322817.1 Intron NP_001309746.1
NM_001322819.1 Intron NP_001309748.1
NM_001322820.1 Intron NP_001309749.1
Gene
WTAP
Gene Name
Wilms tumor 1 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270531.1 Intron NP_001257460.1
NM_001270532.1 Intron NP_001257461.1
NM_001270533.1 Intron NP_001257462.1
NM_004906.4 Intron NP_004897.2
NM_152857.2 Intron NP_690596.1
NM_152858.2 Intron NP_690597.1
XM_017011514.1 Intron XP_016867003.1
XM_017011515.1 Intron XP_016867004.1
XM_017011516.1 Intron XP_016867005.1

View Full Product Details