Product Details

SNP ID
rs9917844
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:94083312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACACAGCAAAGCAAGGAAAGAA[G/T]GCCACAACAGAAGAGATTTATTGAA
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
NSUN3 PubMed Links

Gene Details

Gene
NSUN3
Gene Name
NOP2/Sun RNA methyltransferase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022072.3 Intron NP_071355.1

View Full Product Details