Product Details
- SNP ID
-
rs113994033
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:27366880 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCCGGCTGTCCACCACTACCACC[T/C]GAAACCGCCGGCCCTCTGTCCAAGC
- Phenotype
-
MIM: 606687
MIM: 604883
MIM: 605963
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EIF2B4
PubMed Links
Gene Details
- Gene
- EIF2B4
- Gene Name
- eukaryotic translation initiation factor 2B subunit delta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001034116.1 |
1210 |
Missense Mutation |
CAG,CGG |
Q357R |
NP_001029288.1 |
NM_001318965.1 |
1210 |
Missense Mutation |
CAG,CGG |
Q378R |
NP_001305894.1 |
NM_001318966.1 |
1210 |
Missense Mutation |
CAG,CGG |
Q342R |
NP_001305895.1 |
NM_001318967.1 |
1210 |
Missense Mutation |
CAG,CGG |
Q326R |
NP_001305896.1 |
NM_001318968.1 |
1210 |
Missense Mutation |
CAG,CGG |
Q162R |
NP_001305897.1 |
NM_001318969.1 |
1210 |
Missense Mutation |
CAG,CGG |
Q151R |
NP_001305898.1 |
NM_015636.3 |
1210 |
Missense Mutation |
CAG,CGG |
Q356R |
NP_056451.3 |
NM_172195.3 |
1210 |
Missense Mutation |
CAG,CGG |
Q377R |
NP_751945.2 |
XM_006712132.1 |
1210 |
Missense Mutation |
CAG,CGG |
Q341R |
XP_006712195.1 |
XM_011533147.2 |
1210 |
Missense Mutation |
CAG,CGG |
Q151R |
XP_011531449.1 |
- Gene
- GTF3C2
- Gene Name
- general transcription factor IIIC subunit 2
There are no transcripts associated with this gene.
- Gene
- LOC105374363
- Gene Name
- uncharacterized LOC105374363
There are no transcripts associated with this gene.
- Gene
- SNX17
- Gene Name
- sorting nexin 17
There are no transcripts associated with this gene.
View Full Product Details