Product Details

SNP ID
rs4844212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70145231 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGATTTCAAGCAGAGATATTTCAA[A/G]ATACGTCTCTTGGAAATCTTGATGT
Phenotype
MIM: 300139
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGBP1 PubMed Links

Gene Details

Gene
IGBP1
Gene Name
immunoglobulin (CD79A) binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001551.2 Intron NP_001542.1
XM_017029489.1 Intron XP_016884978.1
XM_017029490.1 Intron XP_016884979.1

View Full Product Details