Product Details

SNP ID
rs10460811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:109329678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGGCTTCATGAAATAAGCTCAG[C/G]GGTCTCAGCTCTTAACACACATTGG
Phenotype
MIM: 614125
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DPPA4 PubMed Links

Gene Details

Gene
DPPA4
Gene Name
developmental pluripotency associated 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018189.3 Intron NP_060659.3
XM_011512954.2 Intron XP_011511256.1

View Full Product Details