Product Details

SNP ID
rs5917575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:38356996 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGCTCCTTTCTTTTGCATGCATT[C/G]TAATATGCCTTTTTGAGAAGGAAAG
Phenotype
MIM: 300461
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OTC PubMed Links

Gene Details

Gene
OTC
Gene Name
ornithine carbamoyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000531.5 Intron NP_000522.3
XM_017029556.1 Intron XP_016885045.1

View Full Product Details