Product Details

SNP ID
rs6810306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122529863 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATAGCATCTAATAAGAGAAGAC[A/G]GAGTGCCTCCTAACCTCATGAACAG
Phenotype
MIM: 612065
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARP9 PubMed Links

Gene Details

Gene
PARP9
Gene Name
poly(ADP-ribose) polymerase family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146102.1 Intron NP_001139574.1
NM_001146103.1 Intron NP_001139575.1
NM_001146104.1 Intron NP_001139576.1
NM_001146105.1 Intron NP_001139577.1
NM_001146106.1 Intron NP_001139578.1
NM_031458.2 Intron NP_113646.2
XM_005247820.2 Intron XP_005247877.1
XM_011513218.1 Intron XP_011511520.1
XM_017007304.1 Intron XP_016862793.1
XM_017007305.1 Intron XP_016862794.1
XM_017007306.1 Intron XP_016862795.1

View Full Product Details