Product Details

SNP ID
rs10156027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102917716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTTAAGCATGTCAAAGACATGA[C/T]AGGAAGATTGTTCCAGGATGACACT
Phenotype
MIM: 609080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM185A PubMed Links

Gene Details

Gene
FAM185A
Gene Name
family with sequence similarity 185 member A
There are no transcripts associated with this gene.

Gene
FBXL13
Gene Name
F-box and leucine rich repeat protein 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111038.1 Intron NP_001104508.1
NM_001287150.1 Intron NP_001274079.1
NM_145032.3 Intron NP_659469.3
XM_005250205.3 Intron XP_005250262.1
XM_005250207.3 Intron XP_005250264.1
XM_005250208.3 Intron XP_005250265.1
XM_005250209.2 Intron XP_005250266.1
XM_006715898.2 Intron XP_006715961.1
XM_011515928.2 Intron XP_011514230.1
XM_011515929.2 Intron XP_011514231.1
XM_011515930.2 Intron XP_011514232.1
XM_011515932.2 Intron XP_011514234.1
XM_017011850.1 Intron XP_016867339.1
XM_017011851.1 Intron XP_016867340.1
XM_017011852.1 Intron XP_016867341.1
XM_017011853.1 Intron XP_016867342.1
Gene
LRRC17
Gene Name
leucine rich repeat containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031692.2 Intron NP_001026862.1
NM_005824.2 Intron NP_005815.2
XM_005250108.1 Intron XP_005250165.1

View Full Product Details