Product Details

SNP ID
rs9411205
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136366011 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAAGGCGCTGGTCTTCACTTCCC[C/T]GTCTGCGGCTCCTCCTTCCCCTTCC
Phenotype
MIM: 607212 MIM: 609491 MIM: 602777
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CARD9 PubMed Links
Additional Information
For this assay, SNP(s) [rs57295785] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CARD9
Gene Name
caspase recruitment domain family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052813.4 Intron NP_434700.2
NM_052814.3 Intron NP_434701.1
Gene
DNLZ
Gene Name
DNL-type zinc finger
There are no transcripts associated with this gene.

Gene
GPSM1
Gene Name
G-protein signaling modulator 1
There are no transcripts associated with this gene.

Gene
SNAPC4
Gene Name
small nuclear RNA activating complex polypeptide 4
There are no transcripts associated with this gene.

View Full Product Details