Product Details

SNP ID
rs9823581
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129284996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCAGCAGCACTATGGAGGGATT[A/G]GCTTTTAAACAAAAGGAAGCCCTTC
Phenotype
MIM: 615525
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
COPG1 PubMed Links
Additional Information
For this assay, SNP(s) [rs60590678,rs60944480] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
COPG1
Gene Name
coatomer protein complex subunit gamma 1
There are no transcripts associated with this gene.

Gene
HMCES
Gene Name
5-hydroxymethylcytosine (hmC) binding, ES cell-specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006109.1 Intron NP_001006109.1
NM_020187.2 Intron NP_064572.2
XM_005247636.3 Intron XP_005247693.1
XM_005247637.3 Intron XP_005247694.1
XM_017006877.1 Intron XP_016862366.1

View Full Product Details