Product Details

SNP ID
rs10019053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:163117227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCATCTGTACCCAAGGTGGAA[C/T]AGACTTTTCTCACCTCTATGTTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAF1 PubMed Links

Gene Details

Gene
NAF1
Gene Name
nuclear assembly factor 1 ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128931.1 Intron NP_001122403.1
NM_138386.2 Intron NP_612395.2
XM_011532410.2 Intron XP_011530712.1
XM_011532411.2 Intron XP_011530713.1

View Full Product Details