Product Details

SNP ID
rs9296145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35307952 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCTGGTGGTGGAAGAAGCCCTTC[A/C]ATATAGCATGTATTGATTGCACTTG
Phenotype
MIM: 610094
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DEF6 PubMed Links

Gene Details

Gene
DEF6
Gene Name
DEF6, guanine nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022047.3 Intron NP_071330.3

View Full Product Details