Product Details

SNP ID
rs9829604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47231095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATGAGGAGCTGGAAGAAACATA[A/T]GATATTTTAAGAGATACAGAGGACA
Phenotype
MIM: 607910
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIF9 PubMed Links

Gene Details

Gene
KIF9
Gene Name
kinesin family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134878.1 Intron NP_001128350.1
NM_022342.4 Intron NP_071737.1
NM_182902.3 Intron NP_878905.2
XM_005265388.4 Intron XP_005265445.1
XM_005265389.4 Intron XP_005265446.1
XM_006713291.3 Intron XP_006713354.1
XM_011534003.2 Intron XP_011532305.1
XM_011534004.2 Intron XP_011532306.1
XM_011534005.2 Intron XP_011532307.1
XM_011534006.2 Intron XP_011532308.1
XM_011534007.2 Intron XP_011532309.1
XM_017007029.1 Intron XP_016862518.1
XM_017007030.1 Intron XP_016862519.1
XM_017007031.1 Intron XP_016862520.1
Gene
KIF9-AS1
Gene Name
KIF9 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details