Product Details

SNP ID
rs10422028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9670678 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTACTAGAGGCCAGAAGGGTAGGA[A/G]GGAGACGGAGATAAAGAGAGGTTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF562 PubMed Links

Gene Details

Gene
ZNF562
Gene Name
zinc finger protein 562
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130031.1 Intron NP_001123503.1
NM_001130032.1 Intron NP_001123504.1
NM_001300885.1 Intron NP_001287814.1
NM_017656.3 Intron NP_060126.2
XM_005259941.3 Intron XP_005259998.1
XM_005259944.3 Intron XP_005260001.1
XM_005259951.3 Intron XP_005260008.1
XM_005259952.3 Intron XP_005260009.1
XM_011528100.2 Intron XP_011526402.1
XM_011528102.2 Intron XP_011526404.1
XM_017026898.1 Intron XP_016882387.1
XM_017026899.1 Intron XP_016882388.1

View Full Product Details