Product Details

SNP ID
rs8179431
Assay Type
Functionally Tested
NCBI dbSNP Submissions
16
Location
Chr.1:156292759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAGTCAGAATGCCTCTCATTTA[A/G]GCCTGCAAGTCCCATCCAAGGGAAA
Phenotype
MIM: 610962 MIM: 615531
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GLMP PubMed Links
Additional Information
For this assay, SNP(s) [rs113301950] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GLMP
Gene Name
glycosylated lysosomal membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256604.1 1209 UTR 3 NP_001243533.1
NM_001256605.1 1209 UTR 3 NP_001243534.1
NM_001256608.1 1209 UTR 3 NP_001243537.1
NM_001256609.1 1209 UTR 3 NP_001243538.1
NM_144580.2 1209 UTR 3 NP_653181.1
XM_011509117.2 1209 UTR 3 XP_011507419.1
XM_017000149.1 1209 UTR 3 XP_016855638.1
Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

Gene
TMEM79
Gene Name
transmembrane protein 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032323.2 1209 Intron NP_115699.1
Gene
VHLL
Gene Name
von Hippel-Lindau tumor suppressor like
There are no transcripts associated with this gene.

View Full Product Details