Product Details

SNP ID
rs10406800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42509608 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGTCAGGATAGTGGTTACTCCT[C/T]TTTGGAGGGAAGTGACTGGTAGAGT
Phenotype
MIM: 109770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEACAM1 PubMed Links

Gene Details

Gene
CEACAM1
Gene Name
carcinoembryonic antigen related cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024912.2 Intron NP_001020083.1
NM_001184813.1 Intron NP_001171742.1
NM_001184815.1 Intron NP_001171744.1
NM_001184816.1 Intron NP_001171745.1
NM_001205344.1 Intron NP_001192273.1
NM_001712.4 Intron NP_001703.2
XM_011527206.1 Intron XP_011525508.1
Gene
LIPE-AS1
Gene Name
LIPE antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details