Product Details
- SNP ID
-
rs10247667
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:123770472 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAAAAAAAAAATCCTGCCGTGGAA[A/G]TTCCAGGGAAGACCTGGGAGAAAAA
- Phenotype
-
MIM: 604510
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HYAL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112385417] are located under a probe and SNP(s) [rs549457728] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HYAL4
- Gene Name
- hyaluronoglucosaminidase 4
View Full Product Details