Product Details

SNP ID
rs9637339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41245278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGCACATACATGGTGTAGGGTA[A/G]GAGGTAAGGAAGGTTGGCTGGGCTG
Phenotype
MIM: 616236 MIM: 602362
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHADL PubMed Links

Gene Details

Gene
CHADL
Gene Name
chondroadherin like
There are no transcripts associated with this gene.

Gene
MIR6889
Gene Name
microRNA 6889
There are no transcripts associated with this gene.

Gene
RANGAP1
Gene Name
Ran GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278651.1 3308 UTR 3 NP_001265580.1
NM_001317930.1 3308 UTR 3 NP_001304859.1
NM_002883.3 3308 UTR 3 NP_002874.1
XM_005261695.1 3308 Intron XP_005261752.1
XM_005261696.1 3308 Intron XP_005261753.1
XM_006724289.3 3308 Intron XP_006724352.1
XM_011530293.1 3308 Intron XP_011528595.1
XM_011530294.2 3308 Intron XP_011528596.1
XM_011530295.2 3308 Intron XP_011528597.1
XM_011530297.1 3308 Intron XP_011528599.1
XM_017028893.1 3308 Intron XP_016884382.1
XM_017028894.1 3308 Intron XP_016884383.1
XM_017028895.1 3308 Intron XP_016884384.1
XM_017028896.1 3308 Intron XP_016884385.1
XM_017028897.1 3308 Intron XP_016884386.1
XM_017028898.1 3308 Intron XP_016884387.1
XM_017028899.1 3308 Intron XP_016884388.1

View Full Product Details