Product Details

SNP ID
rs9636845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30881194 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAGATGCCTCCCAGGGGCTGAC[A/T]TCCACTAGAGACAAAGGTGAGCGGC
Phenotype
MIM: 600064
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP11-1 PubMed Links

Gene Details

Gene
KRTAP11-1
Gene Name
keratin associated protein 11-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175858.2 362 Missense Mutation AGT,TGT S111C NP_787054.1

View Full Product Details