Product Details
- SNP ID
-
rs10135248
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:103655973 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACAAGATGTTTGTCAAATAAGTAC[C/T]GAGGTACACTCTCCTGGACGGGTTC
- Phenotype
-
MIM: 600025
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLC1
PubMed Links
Gene Details
- Gene
- KLC1
- Gene Name
- kinesin light chain 1
View Full Product Details