Product Details

SNP ID
rs6060080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34848281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCATTCATTCTGGTTTTTGCCTA[A/G]GCTGTTCCTCCTGTCCAGAATGTCC
Phenotype
MIM: 612342
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GGT7 PubMed Links

Gene Details

Gene
GGT7
Gene Name
gamma-glutamyltransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178026.2 Intron NP_821158.2
XM_011528782.2 Intron XP_011527084.1
XM_011528783.2 Intron XP_011527085.1
XM_017027809.1 Intron XP_016883298.1
XM_017027810.1 Intron XP_016883299.1
XM_017027811.1 Intron XP_016883300.1

View Full Product Details