Product Details

SNP ID
rs2064044
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:20747572 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGACGTGAGCCACTGCATCTGGC[A/C]AGCTGAAGAAAGTTTTTTCCACTTG
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
LINC00320 PubMed Links

Gene Details

Gene
LINC00320
Gene Name
long intergenic non-protein coding RNA 320
There are no transcripts associated with this gene.

View Full Product Details