Product Details
- SNP ID
-
rs7554961
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
43
- Location
-
Chr.1:223703938 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAACCACACACTCACATCCACACA[C/T]GCCACAGACCCTCTCATGAACATCT
- Phenotype
-
MIM: 114230
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAPN2
PubMed Links
Gene Details
- Gene
- CAPN2
- Gene Name
- calpain 2
View Full Product Details