Product Details

SNP ID
rs9770094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149474807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACCGAAGGGCGCTGCCATCCCG[G/T]GCGCTGCCCCCACCGCTGCCGCCAC
Phenotype
MIM: 613914
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF746 PubMed Links

Gene Details

Gene
ZNF746
Gene Name
zinc finger protein 746
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163474.1 1913 Silent Mutation GCA,GCC A505A NP_001156946.1
NM_152557.4 1913 Silent Mutation GCA,GCC A504A NP_689770.3
XM_005249955.2 1913 Silent Mutation GCA,GCC A520A XP_005250012.1
XM_005249956.2 1913 Silent Mutation GCA,GCC A519A XP_005250013.1
XM_005249957.1 1913 Silent Mutation GCA,GCC A507A XP_005250014.1
XM_011515838.1 1913 Silent Mutation GCA,GCC A492A XP_011514140.1

View Full Product Details