Product Details

SNP ID
rs10521939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:27462870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGAGGTAACTATGTCAGCAATTT[C/G]ATCAAGTGTATTGAGTTCACAGTCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPP4R3CP PubMed Links

Gene Details

Gene
PPP4R3CP
Gene Name
protein phosphatase 4 regulatory subunit 3C, pseudogene
There are no transcripts associated with this gene.

View Full Product Details