Product Details

SNP ID
rs10159325
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:211664789 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTACCACAAATACCTTTACATTT[C/G]TAAGTCAACATTGTCTTACTAATGG
Phenotype
MIM: 604043
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NEK2 PubMed Links

Gene Details

Gene
NEK2
Gene Name
NIMA related kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204182.1 Intron NP_001191111.1
NM_001204183.1 Intron NP_001191112.1
NM_002497.3 Intron NP_002488.1
XM_005273147.1 Intron XP_005273204.1

View Full Product Details