Product Details

SNP ID
rs5762788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:28778782 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTGGGAGGCAGTGGGATGAGAT[A/C]TTTGGATGTCTAATGCCCAGAAGCG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC117 PubMed Links

Gene Details

Gene
CCDC117
Gene Name
coiled-coil domain containing 117
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284263.1 Intron NP_001271192.1
NM_001284264.1 Intron NP_001271193.1
NM_001284265.1 Intron NP_001271194.1
NM_173510.3 Intron NP_775781.1

View Full Product Details