Product Details

SNP ID
rs9819471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100405835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCACCTTGATATCAATTTACTCT[C/T]CCTATAGCATAACTGTAATTTCACT
Phenotype
MIM: 606081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LNP1 PubMed Links

Gene Details

Gene
LNP1
Gene Name
leukemia NUP98 fusion partner 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085451.1 Intron NP_001078920.1
Gene
TOMM70
Gene Name
translocase of outer mitochondrial membrane 70
There are no transcripts associated with this gene.

View Full Product Details