Product Details
- SNP ID
-
rs6093966
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:44359432 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAAGCAAAACTTCCCCAGAAACCT[C/T]CTGGCAGACTCCCCTAGTTCAAGGA
- Phenotype
-
MIM: 600281
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HNF4A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60692239] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HNF4A
- Gene Name
- hepatocyte nuclear factor 4 alpha
- Gene
- R3HDML
- Gene Name
- R3H domain containing like
There are no transcripts associated with this gene.
View Full Product Details