Product Details

SNP ID
rs10157255
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:192325234 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGAACAGGAGGTATTTGGCTTTC[C/T]GTCTCTGAATTAATTTGCTTATGAT
Phenotype
MIM: 612407
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGS21 PubMed Links

Gene Details

Gene
RGS21
Gene Name
regulator of G-protein signaling 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039152.3 Intron NP_001034241.1

View Full Product Details