Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001207025.2 | 4683 | UTR 3 | NP_001193954.1 | ||
NM_001207026.1 | 4683 | Intron | NP_001193955.1 | ||
NM_001247994.1 | 4683 | UTR 3 | NP_001234923.1 | ||
NM_002698.4 | 4683 | UTR 3 | NP_002689.1 | ||
XM_005259010.3 | 4683 | UTR 3 | XP_005259067.1 | ||
XM_011527040.2 | 4683 | UTR 3 | XP_011525342.1 | ||
XM_011527041.2 | 4683 | UTR 3 | XP_011525343.1 | ||
XM_011527042.2 | 4683 | UTR 3 | XP_011525344.1 | ||
XM_011527043.2 | 4683 | UTR 3 | XP_011525345.2 | ||
XM_017026884.1 | 4683 | UTR 3 | XP_016882373.1 | ||
XM_017026885.1 | 4683 | UTR 3 | XP_016882374.1 | ||
XM_017026886.1 | 4683 | UTR 3 | XP_016882375.1 | ||
XM_017026887.1 | 4683 | UTR 3 | XP_016882376.1 | ||
XM_017026888.1 | 4683 | UTR 3 | XP_016882377.1 | ||
XM_017026889.1 | 4683 | UTR 3 | XP_016882378.1 | ||
XM_017026890.1 | 4683 | UTR 3 | XP_016882379.1 | ||
XM_017026891.1 | 4683 | UTR 3 | XP_016882380.1 | ||
XM_017026892.1 | 4683 | UTR 3 | XP_016882381.1 | ||
XM_017026893.1 | 4683 | UTR 3 | XP_016882382.1 | ||
XM_017026894.1 | 4683 | UTR 3 | XP_016882383.1 | ||
XM_017026895.1 | 4683 | UTR 3 | XP_016882384.1 | ||
XM_017026896.1 | 4683 | UTR 3 | XP_016882385.1 |