Product Details
- SNP ID
-
rs6003971
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:20379205 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAACACTACTGCCAACAAAAGTAAT[G/T]AGAGAATATAGATGTTAAATCTATT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
USP41
PubMed Links
Gene Details
- Gene
- USP41
- Gene Name
- ubiquitin specific peptidase 41
View Full Product Details