Product Details

SNP ID
rs10084622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:18274532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAATGTCACACAGGTCAAGAGC[C/T]ATTGACACTTCAAATTTATTGCAAT
Phenotype
MIM: 607247 MIM: 606635
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHODL PubMed Links

Gene Details

Gene
CHODL
Gene Name
chondrolectin
There are no transcripts associated with this gene.

Gene
TMPRSS15
Gene Name
transmembrane protease, serine 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002772.2 Intron NP_002763.2
XM_011529654.1 Intron XP_011527956.1
XM_011529655.1 Intron XP_011527957.1
XM_011529656.2 Intron XP_011527958.1
XM_011529657.1 Intron XP_011527959.1
XM_011529658.1 Intron XP_011527960.1
XM_011529659.1 Intron XP_011527961.1

View Full Product Details