Product Details

SNP ID
rs9910614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2309336 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATGTCTCTTCTGTTCCTATAATG[A/C]ACATGTATGGAATCATCTCCCCAGC
Phenotype
MIM: 610963 MIM: 606477
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMG6 PubMed Links

Gene Details

Gene
SMG6
Gene Name
SMG6, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

Gene
SRR
Gene Name
serine racemase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304803.1 Intron NP_001291732.1
NM_021947.2 Intron NP_068766.1
XM_006721565.3 Intron XP_006721628.1
XM_006721566.3 Intron XP_006721629.1
XM_011523974.2 Intron XP_011522276.1

View Full Product Details