Product Details

SNP ID
rs10037271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139851503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCATATGAAAAATGGAGATGA[G/T]GCTCTTTGGAGTGTTTCTGAGGGGC
Phenotype
MIM: 603818
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NRG2 PubMed Links

Gene Details

Gene
NRG2
Gene Name
neuregulin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184935.1 Intron NP_001171864.1
NM_004883.2 Intron NP_004874.1
NM_013981.3 Intron NP_053584.1
NM_013982.2 Intron NP_053585.1
NM_013983.2 Intron NP_053586.1
XM_005268533.3 Intron XP_005268590.1
XM_006714810.3 Intron XP_006714873.1
XM_011537713.2 Intron XP_011536015.1
XM_017010064.1 Intron XP_016865553.1
XM_017010065.1 Intron XP_016865554.1
XM_017010066.1 Intron XP_016865555.1
Gene
PSD2
Gene Name
pleckstrin and Sec7 domain containing 2
There are no transcripts associated with this gene.

View Full Product Details