Product Details

SNP ID
rs9811343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122325845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTACAAACTTCATACACTTTTGC[C/T]CAAATAGATAGTGTTTTAGTCTTTT
Phenotype
MIM: 184600
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSTA PubMed Links

Gene Details

Gene
CSTA
Gene Name
cystatin A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005213.3 Intron NP_005204.1

View Full Product Details