Product Details

SNP ID
rs5936710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:68669710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCAGTCTGGCTGCAGGCTTGGC[C/G]TCAGCATCAGACTCATCCATCAGCT
Phenotype
MIM: 300689
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STARD8 PubMed Links

Gene Details

Gene
STARD8
Gene Name
StAR related lipid transfer domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142503.2 Intron NP_001135975.1
NM_001142504.2 Intron NP_001135976.1
NM_014725.4 Intron NP_055540.2

View Full Product Details