Product Details
- SNP ID
-
rs9477494
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:17619513 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTAGGAGGAACAGGTTAAAAGACC[C/T]TGTGAGAAAAGTTCTTTCAGTGCTT
- Phenotype
-
MIM: 603948
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM8A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114582845] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM8A1
- Gene Name
- family with sequence similarity 8 member A1
There are no transcripts associated with this gene.
- Gene
- NUP153
- Gene Name
- nucleoporin 153
View Full Product Details