Product Details

SNP ID
rs9872966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:41211850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATTATTGGAAAGGCCTTCTAAC[C/T]GGTGTCCCTGCTTACACCGTTTTCC
Phenotype
MIM: 116806
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNNB1 PubMed Links

Gene Details

Gene
CTNNB1
Gene Name
catenin beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098209.1 Intron NP_001091679.1
NM_001098210.1 Intron NP_001091680.1
NM_001904.3 Intron NP_001895.1
XM_005264886.2 Intron XP_005264943.1
XM_006712983.1 Intron XP_006713046.1
XM_006712984.1 Intron XP_006713047.1
XM_006712985.1 Intron XP_006713048.1
XM_017005738.1 Intron XP_016861227.1

View Full Product Details