Product Details

SNP ID
hCV29830583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62537467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGGAGTTTGCGTGTTTTCTCC[A/T]TGTCTGTGTGGGTTTTCTTAGGGCA
Phenotype
MIM: 608439
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TLK2 PubMed Links

Gene Details

Gene
TLK2
Gene Name
tousled like kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284333.1 Intron NP_001271262.1
NM_001284363.1 Intron NP_001271292.1
NM_006852.3 Intron NP_006843.2

View Full Product Details