Product Details
- SNP ID
-
rs10517201
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:47847749 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTAAAAAGCACAAAGTTAACAGTA[A/T]CAAGAAAAGTACTCTATCATAAGTA
- Phenotype
-
MIM: 605236
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CORIN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114349450] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CORIN
- Gene Name
- corin, serine peptidase
There are no transcripts associated with this gene.
- Gene
- LOC101927179
- Gene Name
- uncharacterized LOC101927179
There are no transcripts associated with this gene.
- Gene
- NFXL1
- Gene Name
- nuclear transcription factor, X-box binding like 1
View Full Product Details