Product Details

SNP ID
rs8104799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19040883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTCTCATCTGTAGCTACCAGGCA[C/T]CCCTTAGGGTTTGGAAGCTTCCTTG
Phenotype
MIM: 607993
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARMC6 PubMed Links

Gene Details

Gene
ARMC6
Gene Name
armadillo repeat containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199196.1 Intron NP_001186125.1
NM_033415.3 Intron NP_219483.1
XM_005260157.3 Intron XP_005260214.1
XM_017027484.1 Intron XP_016882973.1
XM_017027485.1 Intron XP_016882974.1
Gene
SUGP2
Gene Name
SURP and G-patch domain containing 2
There are no transcripts associated with this gene.

View Full Product Details