Product Details

SNP ID
rs6053878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:6076271 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTTCTCTCCCTGACCAGTTGGGA[C/T]AGACACCTGACTGGAATCCTTGACA
Phenotype
MIM: 607900
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FERMT1 PubMed Links

Gene Details

Gene
FERMT1
Gene Name
fermitin family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017671.4 3725 UTR 3 NP_060141.3

View Full Product Details