Product Details

SNP ID
rs10484615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36448126 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGAACATTACTAAAATTTTGGAA[A/G]GCATTGGAATGTAGTAGAAAGGTCC
Phenotype
MIM: 615932
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD20 PubMed Links

Gene Details

Gene
KCTD20
Gene Name
potassium channel tetramerization domain containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286579.1 Intron NP_001273508.1
NM_001286580.1 Intron NP_001273509.1
NM_173562.4 Intron NP_775833.2
XM_005248940.4 Intron XP_005248997.1
XM_006715023.2 Intron XP_006715086.1
XM_011514398.2 Intron XP_011512700.1
Gene
PXT1
Gene Name
peroxisomal, testis specific 1
There are no transcripts associated with this gene.

View Full Product Details