Product Details

SNP ID
rs9910790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21123445 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACCATCTACACATTCCTCCAAGA[A/G]TAAGTATATTTTTTCCCCAAGATAT
Phenotype
MIM: 616160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHRS7B PubMed Links

Gene Details

Gene
DHRS7B
Gene Name
dehydrogenase/reductase 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015510.4 Intron NP_056325.2
XM_005256595.4 Intron XP_005256652.2
XM_005256596.3 Intron XP_005256653.1
XM_011523786.2 Intron XP_011522088.1
XM_017024425.1 Intron XP_016879914.1
XM_017024426.1 Intron XP_016879915.1

View Full Product Details