Product Details
- SNP ID
-
rs7744363
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:36456888 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGTGTGTTGTCCAGGCTAGTCTC[A/C]TGCAATCCTCCTCTCATGCAAACCT
- Phenotype
-
MIM: 615932
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KCTD20
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4713977] are located under a probe and SNP(s) [rs114588005] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCTD20
- Gene Name
- potassium channel tetramerization domain containing 20
View Full Product Details